Disposable Distillation Micro Tubes, Environmental Express
Supplier: ENVIRONMENTAL EXPRESS
C8100A C8100C C8100P
76436-862PK 2263.3 CAD
76436-862 76436-864 76436-866
Disposable Distillation Micro Tubes, Environmental Express
Environmental Test Kits
Save time and improve safety in your laboratory.
  • Eliminate broken glassware—made of polypropylene with hydrophobic membrane and cap
  • No more having to wash glass tubes—just use and toss!
  • Molded-in graduations at 6- and 10-mL for easy measuring
  • Score-line for quick separation of collection tube
  • Compatible with most micro digestion systems
Micro tubes scale down sample and reagent volumes to less than 10 ml for rapid distillations of cyanide, phenols, and ammonia. These user-filled polypropylene tubes work with most micro distillation (Lachat) systems.
The polypropylene tubes consist of two key pieces with a hydrophobic membrane. Choose a tube for cyanide, ammonia, or phenol. Tubes have one press-fit joint that will never leak—and are simply disposed of after use.
Order Now
SPECIFICATIONS
Environmentally Preferable