Snip & Pour™ Oil and Grease Standards, Environmental Express®
Supplier: ENVIRONMENTAL EXPRESS
G2018
G3004
G3021
G3022
G3023
G3024
G3025
G3025-G34
G3026
76436-960PK
281.24
CAD
76436-960
76436-962
76436-964
76436-966
76436-968
76436-970
76436-972
76436-974
76436-976
Snip & Pour™ Oil and Grease Standards, Environmental Express®
Standards
Petroleum Industry Standards
Premeasured controls for oil and grease analysis.
- Meets requirements for EPA Method 1664
- Each lot is quality checked gravitmetrically and by GC
Just snip the top of the PTFE tube and pour the contents into the sample container or extraction apparatus. HEM contents will not adhere to the sides of the single-use tube. Inexpensive Snip & Pour tubes eliminate pipets and require no warming prior to use.
Caution: MDL = method detection limit.