Universal Solids Standard, Environmental Express®
Supplier: ENVIRONMENTAL EXPRESS
F95005 F95005UH
76436-954PK 148.52 CAD
76436-954 76436-956
Universal Solids Standard, Environmental Express®
Standards Environmental Analysis Standards
Universal Solids Standard (USS) provides known QC data for all solids procedures.
  • Obtain batch level quality analysis and control for all solids testing at an affordable price
  • Quality guaranteed—NIST-traceability is shown on the Certificate of Analysis and each parameter has an acceptability range of ±3 standard deviations
  • Choose from three ranges to best fit your testing needs
This product complies with the following methods: SM 2540,ASTM D5907
These Environmental Express universal solids standards cover multiple EPA test method standard control requirements including TS, TSS, VS, and VSS. Use is easy—simply add the bottle contents into your graduated cylinder and fill to volume, or use as a Matrix Spike by adding a bottle directly to your sample.
Certifications: Certificate of Analysis with traceability to NIST, SDS, and dilution instructions.
Order Now
SPECIFICATIONS
Environmentally Preferable