Quantofix Sulfite Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91306
10217-560EA 100.22 CAD
10217-560
Quantofix Sulfite Test Strips, CTL Scientific Supply
Environmental Test Kits
Semi-quantitative test strips for the determination of Sulfite. Ideal for rapid and easy analysis of Sulfite directly on-site. Can be used with test strip reader QUANTOFIX Relax.
Ordering information: Packaged as a box of 100.