COD Standards, Environmental Express®
Supplier: ENVIRONMENTAL EXPRESS
B1030
B1031
76436-808EA
61.62
CAD
76436-808
76436-810
COD Standards, Environmental Express®
Standards
Environmental Analysis Standards
Controls approved for use in both EPA-compliant and mercury-free vials.
- Made from primary standard grade potassium hydrogen phthalate
- Delivered with an SDS and minimum 8 month shelf life (refrigeration required)
- Easy-to-follow dilution directions printed directly on the label
Certifications: This product complies with the following methods: EPA 410.4, SM 5220D-2011, HACH 8000, ASTM D1252-06, ISO 15705:2002, USGS I-3561-06, BS 6068-2.34:1988, DIN 38414-9, and ISO 6060:1989.