Crusher/Splitter Hoods
Supplier: MONASHEE MANUFACTURING CA
107239-14L 107239-14R 107239-15L 107239-15R
CA11022-174EA 8304.47 CAD
CA11022-174 CA11022-176 CA11022-178 CA11022-180 CA11022-182
Crusher/Splitter Hoods
Fume Hoods and Enclosures Ductless Fume Hoods
The crusher/splitter hoods effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants. Hoods can have the option to have your crusher placed on the left- or right-hand side to provide a convenient work area for a sample splitter. Designed for mining applications, our crusher/splitter hoods have a 10" flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).
  • Solid steel construction with powder coating for long lasting finish
  • 10" flange to attach to ducting
  • LED lighting
  • Compressed air hose (air compressor sold separately)
  • Crusher can be positioned on either the right or left-hand side
Crusher/splitter hoods are made of steel and blue powder coated for a long-lasting finish (additional colors available on request). Custom sizes are also available.