Sulfur Biodiesel Standard, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
BFM-10Y BFM-20Y BFM-5Y BFS-100Y BFS-10Y BFS-15Y BFS-20Y BFS-25Y BFS-50Y BFS-5Y
SXBFM-10YEA 388.81 CAD
SXBFM-10Y SXBFM-20Y SXBFM-5Y SXBFS-100Y SXBFS-10Y SXBFS-15Y SXBFS-20Y SXBFS-25Y SXBFS-50Y SXBFS-5Y
Sulfur Biodiesel Standard, SPEX CertiPrep
Standards Petroleum Industry Standards
B100 Biodiesel Standards
Governments worldwide have passed regulations that mandate lower levels of sulfur in biodiesel fuel. To comply with the implementation of these regulations, SPEX CertiPrep offers specifically designed Certified Reference Materials for industrial use.
Certifications: The B100 Biodiesel Standards meet the requirements for testing ASTM Methods D6751, D5453 and EN 14214.