B100 Biodiesel Certified Matrix Blank, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
BF-BLKX BF-BLKY
SXBF-BLKXEA 317.58 CAD
SXBF-BLKX SXBF-BLKY
B100 Biodiesel Certified Matrix Blank, SPEX CertiPrep
Standards Petroleum Industry Standards
B100 Biodiesel Standards
Governments worldwide have passed regulations that mandate lower levels of sulfur in biodiesel fuel. To comply with the implementation of these regulations, SPEX CertiPrep offers specifically designed Certified Reference Materials for industrial use. Our B100 Biodiesel Standards meet the requirements for testing ASTM Methods D6751, D5453 and EN 14214.