Assurance® Grade Selenium +4, Speciation Standard, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
SPEC-SE4 SPEC-SE4M
SXSPEC-SE4EA 115.36 CAD
SXSPEC-SE4 SXSPEC-SE4M
Assurance® Grade Selenium +4, Speciation Standard, SPEX CertiPrep
Standards Cationic Standards Selenium Standards
Speciation analysis has become common in many fields, including environmental, food and pharmaceutical testing labs. To analyze species within a sample requires Certified Reference Materials (CRMs) for sample verification and method validation.
Many speciation standards are available in today’s market, however, most of them are not certified or analyzed with a state-of-the-art ICP, ICP-MS or LC-ICP-MS. SPEX CertiPrep offers a wide variety of speciation standards, certified to the strictest ISO 17034 guidelines, and tested on LC-ICP-MS.