Multi-Element Organometallic Oil Standards, 23 Elements, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
S23-100Y S23-100Z S23-300Y S23-300Z S23-500Y S23-500Z S23-900Y S23-900Z
SXS23-100YEA 439.15 CAD
SXS23-100Y SXS23-100Z SXS23-300Y SXS23-300Z SXS23-500Y SXS23-500Z SXS23-900Y SXS23-900Z
Multi-Element Organometallic Oil Standards, 23 Elements, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
SPEX CertiPrep presents a comprehensive offering of Organometallic Oil Standards. The applications of these standards include:
  • Crude oils
  • Additive metals
  • Environmental monitoring
  • Petrochemical testing
  • Pharmaceuticals
  • Food processing
  • Sulfur in diesel fuel
The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. The examples are endless. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.
- Choice of over 35 single-elements at 1,000 or 5,000 μg/g
- Popular multi-element blends of 23, 21, 12, or 5 elements
- Clear, transparent matrix
- Guaranteed stable and accurate
Certificate of Analysis with every solution
Certifications: Manufactured under an internationally accredited ISO 9001 quality system and compliant with ISO/IEC 17025.