Multi-Element Organometallic Oil Standards, 5 Elements, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
SPEX CertiPrep presents a comprehensive offering of Organometallic Oil Standards. The applications of these standards include:
- Crude oils
- Additive metals
- Environmental monitoring
- Petrochemical testing
- Pharmaceuticals
- Food processing
- Sulfur in diesel fuel
The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. The examples are endless. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.
- Choice of over 35 single-elements at 1,000 or 5,000 μg/g
- Popular multi-element blends of 23, 21, 12, or 5 elements
- Clear, transparent matrix
- Guaranteed stable and accurate
Certificate of Analysis with every solution
Certifications: Manufactured under an internationally accredited ISO 9001 quality system and compliant with ISO/IEC 17025.