Matrix Orbital Delta Thermoshakers
Supplier: IKA Works
0020025648 0020025658 0020025678 0020025668
76417-514EA 6445.94 CAD
76417-514 76417-516 76417-518 76417-520
Matrix Orbital Delta Thermoshakers
Shakers Shaking Incubators
The IKA matrix orbital delta family of thermoshakers with mixing and heating have numerous user-friendly features.
  • Large and clear display with a convenient menu structure allows for intuitive operation
  • Speeds of up to 2000 min⁻¹ (1500 for the Δ F1.5 and Δ F2.0 models) and shaking diameter of 3 mm ensure the liquid mixes quickly and effectively
  • Two different speed modes for gentle heating of the samples
  • Programmable ramps allow for automatic execution of mixing and heating tasks
  • Smart speed adjustment according to insert, attachment and filling volume
  • Aluminium die cast housing ensures secure positioning, excellent stability and everyday robustness
  • RS232 and USB interfaces
Whether it is blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays – samples with even the smallest volumes mix reliably and completely in all laboratory applications. No cross-contamination and an optimal mixing result.
IKA Matrix Orbital Δ FP mixes, heats and has a fixed universal holder for microtiter plates, it has a secure, quick lock for fast and easy change of inserts and attachments as well as automatic attachment detection.
IKA Matrix Orbital Δ F2.0 mixes and heats with a fixed insert for 24×2,0 ml tubes.
IKA Matrix Orbital Δ F1.5 mixes and heats with a fixed insert for 24×1,5 ml tubes.
IKA Matrix Orbital Δ F0.5 mixes and heats with a fixed insert for 24×0,5 ml tubes.
Certifications: IP Protection Class according to DIN EN 60529: IP 21
Ordering information: Supplied without accessories, which must be ordered separately. There is a wide range of accessories available for the IKA Matrix series that allow the optimal operation for your specific application.