HotBlock® Digestion Systems, Environmental Express®
Supplier: Environmental Express
C6002 C6002-240 SC100 SC100-240 SC150 SC150-240 SC151 SC151-240 SC154 SC154-240 SC196 SC196-240
76436-820EA 8894.39 CAD
76436-820 76436-822 76437-064 76437-066 76437-068 76437-070 76437-072 76437-074 76437-076 76437-078 76437-084 76437-086
HotBlock® Digestion Systems, Environmental Express®
Digestion Systems Mineralisation Workstations
EPA-approved systems that simplify and improve metals prep and digestions.
  • Blocks are PTFE coated for chemical and corrosion resistance
  • Integrated block heater and controller design
  • Set high and low temperature alarm limits
  • Include polycarbonate transfer racks designed to set on the block
The HotBlock® Systems provide faster, cleaner, and more cost-effective metals digestions, and are accepted for EPA Methods. Block heaters have an integrated controller and mat/block design and come in configurations to work directly with the HotBlock digestion cupsMore Product Information