Avantor® Hichrom, Retention Gap/Precolumn, HMDS Deactivated Fused Silica
Supplier: VWR International
HI96-05-D-1 HI96-05-D-2 HI96-05-D-5 HI96-05-D-10 HI96-10-D-1 HI96-10-D-2 HI96-10-D-5 HI96-10-D-10 HI96-10-D-20 HI96-15-D-1 HI96-15-D-2 HI96-15-D-5 HI96-15-D-10 HI96-15-D-20 HI96-15-D-30 HI96-18-D-1 HI96-18-D-2 HI96-18-D-5 HI96-18-D-10 HI96-18-D-20 HI96-18-D-30 HI96-20-D-1 HI96-20-D-2 HI96-20-D-5 HI96-20-D-10 HI96-20-D-20 HI96-20-D-30 HI96-25-D-1 HI96-25-D-2 HI96-25-D-5 HI96-25-D-10 HI96-25-D-20 HI96-25-D-30 HI96-25-D-60 HI96-32-D-1 HI96-32-D-2 HI96-32-D-5 HI96-32-D-10 HI96-32-D-20 HI96-32-D-30 HI96-32-D-60 HI96-53-D-1 HI96-53-D-2 HI96-53-D-5 HI96-53-D-10 HI96-53-D-20 HI96-53-D-30 HI96-53-D-60
76436-108EA 62.47 CAD
76436-108 76436-110 76436-112 76436-114 76436-116 76436-118 76436-120 76436-122 76436-124 76436-126 76436-128 76436-130 76436-132 76436-134 76436-136 76436-138 76436-140 76436-142 76436-144 76436-146 76436-148 76436-150 76436-152 76436-154 76436-156 76436-158 76436-160 76436-162 76436-164 76436-166 76436-168 76436-170 76436-172 76436-174 76436-176 76436-178 76436-180 76436-182 76436-184 76436-186 76436-188 76436-190 76436-192 76436-194 76436-196 76436-198 76436-200 76436-202
Avantor® Hichrom, Retention Gap/Precolumn, HMDS Deactivated Fused Silica
Chromatography Columns
HMDS deactivated fused silica retention gap / precolumn. Deactivation mainly for apolar solvents.