Avantor® Hichrom HI-8 HT, GC Columns
Supplier: VWR International
76431-292 76431-676 76431-678 76431-680 76431-682 76431-684 76431-686 76431-688 76431-690 76431-692 76431-694 76431-696 76431-698 76431-700 76431-702 76431-704 76431-706 76431-708 76431-710 76431-712 76431-714 76431-716 76431-822 76431-824
76431-292EA 2218.21 CAD
76431-292 76431-676 76431-678 76431-680 76431-682 76431-684 76431-686 76431-688 76431-690 76431-692 76431-694 76431-696 76431-698 76431-700 76431-702 76431-704 76431-706 76431-708 76431-710 76431-712 76431-714 76431-716 76431-822 76431-824
Avantor® Hichrom HI-8 HT, GC Columns
Chromatography Columns
High Temperature general purpose column ideal for PCBs compounds, Pesticides, environmental analysis.
Maximum temperature limits may change depending on the film thickness.