Avantor® Hichrom HI-DEX DMT Beta, GC Columns
Supplier: VWR International
76429-994 76429-996 76429-998 76430-000 76430-002 76430-004 76430-006 76430-008 76430-010 76430-012 76430-082 76430-084
76429-994EA 2449.41 CAD
76429-994 76429-996 76429-998 76430-000 76430-002 76430-004 76430-006 76430-008 76430-010 76430-012 76430-082 76430-084
Avantor® Hichrom HI-DEX DMT Beta, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.