Avantor® Hichrom HI-XMLB, GC Columns
Supplier: VWR International
HI20-18-018-20 HI20-18-018-30 HI20-20-033-25 HI20-25-010-15 HI20-25-010-30 HI20-25-015-30 HI20-25-015-60 HI20-25-025-15 HI20-25-025-30 HI20-25-025-60 HI20-25-050-30 HI20-25-100-30 HI20-32-025-30 HI20-32-025-60 HI20-32-050-30 HI20-32-050-60 HI20-53-150-15 HI20-53-150-30
76429-818EA 3046.9 CAD
76429-818 76429-820 76429-822 76429-824 76429-826 76429-828 76429-830 76429-832 76429-834 76429-836 76429-838 76429-840 76429-842 76429-844 76429-846 76429-848 76429-850 76429-852
Avantor® Hichrom HI-XMLB, GC Columns
Chromatography Columns
Proprietary low-polarity and low-bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
  • Phase composition: crossbonded low-polarity proprietary phase
  • Long lifetime
Max temperature limits may change depending on the film thickness.