Avantor® Hichrom HI-10, GC Columns
Supplier: VWR International
HI2620010100 HI2620010105 HI26-20-010-40 HI26-20-010-50 HI26-20-010-60 HI2620020100 HI2620020105 HI26-20-020-40 HI26-20-020-50 HI26-20-020-60 HI2625020100 HI2625020105 HI26-25-020-30 HI26-25-020-50 HI26-25-020-60 HI2625025100 HI2625025105 HI26-25-025-30 HI26-25-025-50 HI26-25-025-60 HI2632020100 HI2632020105 HI26-32-020-30 HI26-32-020-50 HI26-32-020-60 HI2632025100 HI2632025105 HI26-32-025-30 HI26-32-025-50 HI26-32-025-60 HI26-18-010-10 HI26-18-010-20 HI26-20-010-10 HI26-20-010-20 HI26-20-020-20
76424-380EA 551.47 CAD
76424-380 76424-382 76424-384 76424-386 76424-388 76424-390 76424-392 76424-394 76424-396 76424-398 76424-400 76424-402 76424-404 76424-406 76424-408 76424-410 76424-412 76424-414 76424-416 76424-418 76424-420 76424-422 76424-424 76424-426 76424-428 76424-430 76424-432 76424-434 76424-436 76424-438 76424-440 76430-128 76430-130 76430-132 76430-134 76430-136
Avantor® Hichrom HI-10, GC Columns
Chromatography Columns
Application specific highly polar phase, ideal for detailed cis/trans FAMEs separation.
  • Phase composition: 100% cyanopropyl polysiloxane
Max temperature limits may change depending on the film thickness.