Avantor® Hichrom HI-5 HT, GC Columns
Supplier: VWR International
HI873201010G2 HI87-25-010-15 HI87-25-010-30 HI87-25-015-15 HI87-25-015-30 HI87-25-025-15 HI87-25-025-30 HI87-32-010-10 HI87-32-010-15 HI87-32-010-30 HI87-32-015-15 HI87-32-015-30 HI87-32-025-15 HI87-32-025-30 HI87-53-010-10 HI87-53-010-15 HI87-53-010-30 HI87-53-015-15 HI87-53-015-30 HI87-53-017-30 HI87-53-025-15 HI87-53-025-30 HI87-10-010-10 HI87-10-010-5
76431-186EA 1511.81 CAD
76431-186 76431-624 76431-626 76431-628 76431-630 76431-632 76431-634 76431-636 76431-638 76431-640 76431-642 76431-644 76431-646 76431-648 76431-650 76431-652 76431-654 76431-656 76431-658 76431-660 76431-662 76431-664 76431-818 76431-820
Avantor® Hichrom HI-5 HT, GC Columns
Chromatography Columns
General purpose column with added temperature stability. For the analysis of solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, pesticides, phenols and more.
Max temperature limits may change depending on the film thickness.