Avantor® Hichrom HI-DAI 1, GC Columns
Supplier: VWR International
76426-562 76426-564 76426-566 76426-568 76426-570 76426-572
76426-562EA 363.84 CAD
76426-562 76426-564 76426-566 76426-568 76426-570 76426-572
Avantor® Hichrom HI-DAI 1, GC Columns
Chromatography Columns
Application specifc column for direct introduction of aqueous samples, thus minimizing sample preparation.
Max temperature limits may change depending on the film thickness.
Order Now
SPECIFICATIONS
Phase name HI-DAI 1
Environmentally Preferable
Phase composition Proprietary Phase developed for direct aqueous injections
Crossbond Yes
Temperature limit 320 °C