Avantor® Hichrom HI-DAI 2, GC Columns
Supplier: VWR International
76426-574 76426-576 76426-578 76426-580 76426-582 76426-584
76426-574EA 3089.71 CAD
76426-574 76426-576 76426-578 76426-580 76426-582 76426-584
Avantor® Hichrom HI-DAI 2, GC Columns
Chromatography Columns
Application specifc column for direct introduction of aqueous samples, thus minimizing sample preparation.
Max temperature limits may change depending on the film thickness.