Avantor® Hichrom HI-DEX B-SE, GC Columns
Supplier: VWR International
76429-874 76429-876 76429-878 76429-880 76429-882 76429-884 76429-886 76429-888 76429-890 76429-892 76430-058 76430-060
76429-874EA 3529.77 CAD
76429-874 76429-876 76429-878 76429-880 76429-882 76429-884 76429-886 76429-888 76429-890 76429-892 76430-058 76430-060
Avantor® Hichrom HI-DEX B-SE, GC Columns
Chromatography Columns
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Order Now
SPECIFICATIONS
Phase name HI-DEX B-SE
Environmentally Preferable
Phase composition Proprietary Cyclodexstrin based
Polarity Chiral stationary phase
Crossbond No
Temperature limit 230 °C