Avantor® Hichrom HI-VOC 1, GC Columns
Supplier: VWR International
76429-110 76429-112 76429-114 76429-116 76429-118 76429-120 76429-122 76429-124 76429-126 76429-128 76429-130 76429-132 76429-134 76429-136 76429-138 76429-140 76429-142 76429-144 76429-146 76429-148 76429-150 76429-152 76429-154 76429-156 76429-158 76429-160 76429-162 76429-164 76429-166 76429-168 76429-170 76429-172 76429-174 76429-176 76429-178 76429-180 76429-182 76429-184 76429-186 76429-188 76429-190 76429-192 76429-194
76429-110EA 353.87 CAD
76429-110 76429-112 76429-114 76429-116 76429-118 76429-120 76429-122 76429-124 76429-126 76429-128 76429-130 76429-132 76429-134 76429-136 76429-138 76429-140 76429-142 76429-144 76429-146 76429-148 76429-150 76429-152 76429-154 76429-156 76429-158 76429-160 76429-162 76429-164 76429-166 76429-168 76429-170 76429-172 76429-174 76429-176 76429-178 76429-180 76429-182 76429-184 76429-186 76429-188 76429-190 76429-192 76429-194
Avantor® Hichrom HI-VOC 1, GC Columns
Chromatography Columns
Application specifc columns for volatile organic compounds (VOCs), solvents and purgeable compounds. Due to the high max. temperature, they are ideal for two-parallel columns confg. in the same oven. A good alternative to the HI-624 giving different selectivity. It is a more robust than the HI-624 and can operate at higher temperatures.
Max temperature limits may change depending on the film thickness.