Avantor® Hichrom HI-17 HT, GC Columns
Supplier: VWR International
HI90-25-010-15 HI90-25-010-30 HI90-25-015-15 HI90-25-015-30 HI90-25-025-15 HI90-25-025-30 HI90-32-010-10 HI90-32-010-15 HI90-32-010-30 HI90-32-015-15 HI90-32-015-30 HI90-32-025-15 HI90-32-025-30 HI90-53-010-10 HI90-53-010-15 HI90-53-010-30 HI90-53-015-15 HI90-53-015-30 HI90-53-017-30 HI90-53-025-15 HI90-53-025-30 HI90-10-010-10 HI90-10-010-5
76431-498EA 1617.51 CAD
76431-498 76431-500 76431-502 76431-504 76431-506 76431-508 76431-510 76431-512 76431-514 76431-516 76431-518 76431-520 76431-522 76431-524 76431-526 76431-528 76431-530 76431-532 76431-534 76431-536 76431-538 76431-806 76431-808
Avantor® Hichrom HI-17 HT, GC Columns
Chromatography Columns
High Temperature general purpose column. Ideal for confrmation analysis, ideal as high polarity dimension in GCxGC-HT confgurations.
Max temperature limits may change depending on the film thickness.