Avantor® Hichrom HI-DEX G-03, GC Columns
Supplier: VWR International
76430-034 76430-036 76430-038 76430-040 76430-042 76430-044 76430-046 76430-048 76430-050 76430-052 76430-090 76430-092
76430-034EA 3529.77 CAD
76430-034 76430-036 76430-038 76430-040 76430-042 76430-044 76430-046 76430-048 76430-050 76430-052 76430-090 76430-092
Avantor® Hichrom HI-DEX G-03, GC Columns
Chromatography Columns
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.