Avantor® Hichrom HI-1 HT, GC Columns
Supplier: VWR
Avantor® Hichrom
HI86-53-025-15 HI86-53-010-10 HI86-32-025-30 HI86-32-010-30 HI86-53-017-30 HI863201010G2 HI86-10-010-10 HI86-53-015-30 HI86-53-025-30 HI86-53-010-15 HI86-10-010-5 HI86-25-010-30 HI86-25-025-30 HI86-25-015-30 HI86-53-010-30 HI86-32-015-30 HI86-32-025-15 HI86-32-010-10 HI86-53-015-15 HI86-25-010-15 HI86-25-025-15 HI86-25-015-15 HI86-32-015-15 HI86-32-010-15
76431-804EA 1667.06 CAD
76431-804 76431-802 76431-484 76431-462 76431-122 76431-496 76431-474 76431-482 76431-460 76431-494 76431-472 76431-480 76431-492 76431-470 76431-490 76431-468 76431-458 76431-466 76431-488 76431-478 76431-456 76431-486 76431-464 76431-476
Avantor® Hichrom HI-1 HT, GC Columns
Chromatography Columns
General purpose GC-MS column Solvent impurities, PCBs, SimDist, drugs, natural gases, essential oils, semivolatiles, pesticides, phenols, etc.
Max temperature limits may change depending on the film thickness.