Avantor® Hichrom HI-DEX DET Beta, GC Columns
Supplier: VWR International
76429-934 76429-936 76429-938 76429-940 76429-942 76429-944 76429-946 76429-948 76429-950 76429-952 76430-070 76430-072
76429-934EA 403.75 CAD
76429-934 76429-936 76429-938 76429-940 76429-942 76429-944 76429-946 76429-948 76429-950 76429-952 76430-070 76430-072
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.