VapLock™ Solvent Collection Systems with Quick Disconnects and FM-Approved Container, Cole-Parmer®
Supplier: Antyila Scientific
12020-69 12020-70
76441-336EA 1246.89 CAD
76441-336 76441-338
VapLock™ Solvent Collection Systems with Quick Disconnects and FM-Approved Container, Cole-Parmer®
Filters Liquid Process Filters
Complete solution to safely collect solvent waste from laboratory instruments.
  • FM-approved container offers added safety
  • Collect solvent waste from up to 10 effluent streams
  • Minimize exposure to harmful chemical vapors
  • Quick-disconnect fitting material made of stainless steel
  • Translucent bottle
Solvent waste systems come complete with everything you need to safely collect solvent waste from any system that generates solvent waste. Closed-system design reduces harmful and flammable solvent vapor emissions into the environment as well as leaks andMore Product Information