Assurance® Grade Titanium Standards for AA and ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
PLTI5-2X PLTI5-3X PLTI9-2M PLTI9-2T PLTI9-2X PLTI9-2Y PLTI9-3X PLTI9-3Y
SXPLTI5-2XEA 360 CAD
SXPLTI5-2X SXPLTI5-3X SXPLTI9-2M SXPLTI9-2T SXPLTI9-2X SXPLTI9-2Y SXPLTI9-3X SXPLTI9-3Y
Assurance® Grade Titanium Standards for AA and ICP, SPEX CertiPrep
Standards Cationic Standards Titanium Standards
Single-element standards for AA & ICP.
  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)
AA & ICP Assurance® Grade CRMs are designed for AA and ICP and are available in single and multi-element formulations. 70 elements are available as single-element standards and are available at 1,000 µg/ml and/or 10,000 µg/ml. They are available packaged in 30 ml, 125 ml, 250 ml, and 500 ml bottles to minimize contamination.
Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016