Assurance® Grade Indium Standards for AA and ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
PLIN2-2M PLIN2-2X PLIN2-2Y
SXPLIN2-2MEA 69.05 CAD
SXPLIN2-2M SXPLIN2-2X SXPLIN2-2Y
Assurance® Grade Indium Standards for AA and ICP, SPEX CertiPrep
Standards Cationic Standards Indium Standards
Single-element standards for AA & ICP.
  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)
AA & ICP Assurance® Grade CRMs are designed for AA and ICP and are available in single and multi-element formulations. 70 elements are available as single-element standards and are available at 1,000 µg/ml and/or 10,000 µg/ml. They are available packaged in 30 ml, 125 ml, 250 ml, and 500 ml bottles to minimize contamination.
Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016