Assurance® Grade Mercury Standards for AA and ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
PLHG2-1AX PLHG2-1AY PLHG2-1X PLHG2-1Y PLHG4-2M PLHG4-2T PLHG4-2X PLHG4-2Y PLHG4-3X PLHG4-3Y
SXPLHG2-1AXEA 213.49 CAD
SXPLHG2-1AX SXPLHG2-1AY SXPLHG2-1X SXPLHG2-1Y SXPLHG4-2M SXPLHG4-2T SXPLHG4-2X SXPLHG4-2Y SXPLHG4-3X SXPLHG4-3Y
Assurance® Grade Mercury Standards for AA and ICP, SPEX CertiPrep
Standards Cationic Standards Mercury Standards
Single-element standards for AA & ICP.
  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)
AA & ICP Assurance® Grade CRMs are designed for AA and ICP and are available in single and multi-element formulations. 70 elements are available as single-element standards and are available at 1,000 µg/ml and/or 10,000 µg/ml. They are available packaged in 30 ml, 125 ml, 250 ml, and 500 ml bottles to minimize contamination.
Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016