Assurance® Grade Aluminum Standards for AA and ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
PLAL1-2X PLAL1-3X PLAL2-2M PLAL2-2T PLAL2-2X PLAL2-2Y PLAL2-3X PLAL2-3Y
SXPLAL1-2XEA 271.21 CAD
SXPLAL1-2X SXPLAL1-3X SXPLAL2-2M SXPLAL2-2T SXPLAL2-2X SXPLAL2-2Y SXPLAL2-3X SXPLAL2-3Y
Assurance® Grade Aluminum Standards for AA and ICP, SPEX CertiPrep
Standards Cationic Standards Aluminum Standards
Single-element standards for AA & ICP.
  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)
AA & ICP Assurance® Grade CRMs are designed for AA and ICP and are available in single and multi-element formulations. 70 elements are available as single-element standards and are available at 1,000 µg/ml and/or 10,000 µg/ml. They are available packaged in 30 ml, 125 ml, 250 ml, and 500 ml bottles to minimize contamination.
Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016