Assurance® Grade TCLP Standard without Mercury for AA & ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
Assurance® mixed multi-element standards for AA & ICP. Toxicity Characteristic Leachate Performance (TCLP) Standards
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
Designed to determine the mobility of the Inorganic contaminants present in liquid, solid and multi-phase wastes. To simplify, TCLP is designed to determine the hazardous contaminants that are actually entering into the environment. In addition to the SPEX CertiPrep TCLP TCLP Standards, designed with all of the elements in one solution, the Toxicity Characteristic rule separates the elements according to specific instrumentation: ICP, GFAA, and Cold Vapor AA.
For use in accordance with the Toxicity Characteristic Rule Regulatory Levels issues in the Federal Register 55, 11846 March 1990; Method 1311.