Assurance® Grade Set of 2 Multi-Element Quality Control Standards for AA & ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
QC-SETA QC-SETB
SXQC-SETAEA 943.69 CAD
SXQC-SETA SXQC-SETB
Assurance® Grade Set of 2 Multi-Element Quality Control Standards for AA & ICP, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
Assurance® mixed multi-element quality control standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
Quality Control Standards are used to check the standard curve, the procedure for inter-element correction and other spectral interferences. These standards are carried through the entire analytical operation of the method. If the determined concentration is not within +/- 5% of 1 µg/mL, the laboratory performance is unacceptable. The source of the problem should be identified and corrected before continuing the analysis.