Assurance® Grade Mixed Calibration Standard 2A for AA and ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
MIXSTD2A-100 MIXSTD2A-500
SXMIXSTD2A-100EA 316.57 CAD
SXMIXSTD2A-100 SXMIXSTD2A-500
Assurance® Grade Mixed Calibration Standard 2A for AA and ICP, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
Assurance® mixed multi-element standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
AA & ICP Calibration Standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.
For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition).
Order Now
SPECIFICATIONS
Environmentally Preferable