Assurance® Grade Interference Check Standard 5 for AA & ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
INTER5-100 INTER5-500
SXINTER5-100EA 527.67 CAD
SXINTER5-100 SXINTER5-500
Assurance® Grade Interference Check Standard 5 for AA & ICP, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
Assurance® mixed multi-element interference check standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
The Interference Check Standards are used to set or confirm that the correct background correction intervals have been set for sequential ICP spectrometers and that the proper inter-element correction factors are set for simultaneous ICP spectrometers.
For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition).