Assurance® Grade Environmental EPA Set with Mercury and without Mercury for AA & ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
EPA-SET EPA-SETN
SXEPA-SETEA 3427.98 CAD
SXEPA-SET SXEPA-SETN
Assurance® Grade Environmental EPA Set with Mercury and without Mercury for AA & ICP, SPEX CertiPrep
Standards Environmental Analysis Standards
Assurance® mixed multi-element standards for AA & ICP. Interference check standards
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
For use in US EPA Method 6010 and 200.7 (Revision 4.4).