Avantor® Hichrom HI-PS255, GC Columns
Supplier: VWR International
76428-382 76428-384 76428-386 76428-388 76428-390 76428-392 76428-394 76428-396 76428-398 76428-224 76428-226 76428-228 76428-230 76428-232 76428-234 76428-236 76428-238 76428-240 76428-242 76428-244 76428-246 76428-248 76428-250 76428-252 76428-254 76428-256 76428-258 76428-260 76428-262 76428-264 76428-266 76428-268 76428-270 76428-272 76428-274 76428-276 76428-278 76428-280 76428-282 76428-284 76428-286 76428-288 76428-290 76428-292 76428-294 76428-296 76428-298 76428-300 76428-302 76428-304 76428-306 76428-308 76428-310 76428-312 76428-314 76428-316 76428-318 76428-320 76428-322 76428-324 76428-326 76428-328 76428-330 76428-332 76428-334 76428-336 76428-338 76428-340 76428-342 76428-344 76428-346 76428-348 76428-350 76428-352 76428-354 76428-356 76428-358 76428-360 76428-362 76428-364 76428-366 76428-368 76428-370 76428-372 76428-374 76428-376 76428-378 76428-380 76428-400 76428-402 76428-404 76428-406 76428-408 76428-410 76428-412 76428-414 76428-416 76428-418 76428-420 76428-422 76428-424 76428-426 76428-428 76428-430 76428-432 76428-434 76428-436 76428-438 76428-440 76428-442 76428-444 76428-446 76428-448 76428-450 76428-452 76428-454 76428-456 76428-458 76428-460 76428-462 76428-464 76428-466 76428-468 76428-470 76428-472 76428-474 76428-476 76428-478 76428-480 76428-482 76428-484 76428-486 76428-488 76428-490 76428-492 76428-494 76428-496 76428-498 76428-500 76428-502 76428-504 76428-506 76428-508 76428-510 76428-512 76428-514 76428-516 76428-518 76428-520 76430-778 76430-780 76430-782 76430-784 76430-786 76430-788 76430-790 76430-792 76430-794 76430-796 76430-798 76430-800 76430-802 76430-804 76430-806 76430-808 76430-810 76431-358 76431-360 76431-362 76431-364 76431-366
76428-382EA 1439.16 CAD
76428-382 76428-384 76428-386 76428-388 76428-390 76428-392 76428-394 76428-396 76428-398 76428-224 76428-226 76428-228 76428-230 76428-232 76428-234 76428-236 76428-238 76428-240 76428-242 76428-244 76428-246 76428-248 76428-250 76428-252 76428-254 76428-256 76428-258 76428-260 76428-262 76428-264 76428-266 76428-268 76428-270 76428-272 76428-274 76428-276 76428-278 76428-280 76428-282 76428-284 76428-286 76428-288 76428-290 76428-292 76428-294 76428-296 76428-298 76428-300 76428-302 76428-304 76428-306 76428-308 76428-310 76428-312 76428-314 76428-316 76428-318 76428-320 76428-322 76428-324 76428-326 76428-328 76428-330 76428-332 76428-334 76428-336 76428-338 76428-340 76428-342 76428-344 76428-346 76428-348 76428-350 76428-352 76428-354 76428-356 76428-358 76428-360 76428-362 76428-364 76428-366 76428-368 76428-370 76428-372 76428-374 76428-376 76428-378 76428-380 76428-400 76428-402 76428-404 76428-406 76428-408 76428-410 76428-412 76428-414 76428-416 76428-418 76428-420 76428-422 76428-424 76428-426 76428-428 76428-430 76428-432 76428-434 76428-436 76428-438 76428-440 76428-442 76428-444 76428-446 76428-448 76428-450 76428-452 76428-454 76428-456 76428-458 76428-460 76428-462 76428-464 76428-466 76428-468 76428-470 76428-472 76428-474 76428-476 76428-478 76428-480 76428-482 76428-484 76428-486 76428-488 76428-490 76428-492 76428-494 76428-496 76428-498 76428-500 76428-502 76428-504 76428-506 76428-508 76428-510 76428-512 76428-514 76428-516 76428-518 76428-520 76430-778 76430-780 76430-782 76430-784 76430-786 76430-788 76430-790 76430-792 76430-794 76430-796 76430-798 76430-800 76430-802 76430-804 76430-806 76430-808 76430-810 76431-358 76431-360 76431-362 76431-364 76431-366
Avantor® Hichrom HI-PS255, GC Columns
Chromatography Columns
Apolar phase, suitable for high flm thickness columns, to analyze solvents, alcohols, volatiles, and so on.
Max temperature limits may change depending on the film thickness.