Avantor® Hichrom HI-5, GC Columns
Supplier: VWR
Avantor® Hichrom
HI02-32-100-10 HI02-10-010-10 HI02-53-100-60 HI02-32-050-60 HI02-32-150-30 HI02-20-040-60 HI0232300105 HI02-32-150-60 HI0232025105 HI02-53-500-50 HI02-25-050-60 HI02-32-300-15 HI02-53-500-75 HI02-20-100-40 HI02-53-050-60 HI02-32-025-30 HI02-32-150-10 HI02-32-150-15 HI02-18-040-40 HI02-53-100-10 HI02-18-040-60 HI02-18-018-20 HI02-53-300-50 HI02-53-010-15 HI02-53-700-30 HI02-53-150-10 HI02-15-015-5 HI02-20-010-20 HI025305030G10 HI02-25-025-30 HI02-53-300-75 HI02-25-010-50 HI02-18-040-50 HI02-53-700-15 HI02-32-300-50 HI02-53-050-30 HI02-32-100-30 HI0225025105 HI02-20-010-10 HI02-05-005-2 HI02-53-025-30 HI022502560G5 HI02-32-050-15 HI02-32-025-15 HI02-32-100-50 HI02-32-300-10 HI02-53-500-15 HI02-53-100-30 HI02-05-005-5 HI02-32-500-60 HI02-20-040-10 HI02-15-015-20 HI02-53-150-60 HI02-10-020-5 HI02-53-500-30 HI02-53-050-50 HI02-20-100-10 HI02-25-100-15 HI02-10-020-15 HI023210030G5 HI02-25-025-60 HI02-53-025-15 HI02-20-040-50 HI02-53-300-15 HI02-53-050-15 HI02-53-025-50 HI022510030G5 HI02-25-050-50 HI0232100105 HI02-53-100-75 HI02-32-010-10 HI02-53-010-50 HI02-53-050-10 HI02-53-150-75 HI02-20-020-60 HI02-32-050-50 HI02-32-010-15 HI02-18-040-10 HI02-25-010-15 HI02-20-010-60 HI0232150105 HI02-20-020-40 HI02-18-018-60 HI02-25-010-30 HI02-25-025-15 HI02-32-050-30 HI02-53-265-10 HI02-53-500-10 HI02-20-100-20 HI0225100105 HI02-53-010-30 HI02-25-050-10 HI02-18-020-50 HI02-18-040-20 HI0225050105 HI02-25-010-60 HI02-32-500-10 HI022502530G5 HI02-53-150-15 HI02-25-050-30 HI02-10-040-10 HI02-32-025-60 HI02-53-150-50 HI02-15-015-10 HI02-32-500-30 HI02-32-100-15 HI02-53-025-60 HI02-10-040-15 HI02-32-300-60 HI02-20-010-40 HI02-18-020-40 HI02-53-300-30 HI0232050105 HI02-18-018-50 HI02-53-100-50 HI02-18-020-20 HI02-53-100-15 HI02-10-010-15 HI02-18-018-40 HI02-20-020-10 HI02050052.5 HI0232500105 HI02-25-100-50 HI02-53-500-60 HI02-15-015-30 HI02-18-018-10 HI02-32-100-60 HI02-10-010-5 HI02-18-020-60 HI02-20-100-50 HI02-20-020-20 HI02-53-300-10 HI02-32-500-50 HI02-53-300-60 HI02-53-150-30 HI02-32-010-60 HI02-32-300-30 HI02-10-040-5 HI02-32-500-15 HI025310030G5 HI02-32-050-10 HI02-20-020-50 HI0232010105 HI02-25-100-60 HI02-25-050-15 HI02-20-100-60 HI02-53-010-10 HI02-25-025-50 HI02-20-010-50 HI022502515G5 HI02-25-100-30 HI02-10-020-10 HI02-20-040-40 HI02-32-400-30 HI02-32-025-50 HI02-32-010-30 HI02-18-020-10 HI02-25-025-10 HI02-53-010-60 HI02-32-150-50 HI02-32-010-50 HI02-20-040-20 HI02-32-025-10 HI02-15-015-15 HI02-25-100-10
76435-026EA 1241.2 CAD
76435-026 76435-148 76435-024 76435-146 76435-022 76435-144 76435-020 76435-142 76435-140 76434-980 76434-982 76430-440 76434-984 76430-442 76434-976 76434-978 76435-138 76435-018 76435-136 76430-432 76435-016 76435-134 76430-434 76435-014 76435-132 76430-436 76435-012 76430-438 76435-130 76435-010 76435-098 76435-096 76434-970 76435-094 76434-972 76435-092 76430-430 76435-090 76434-974 76434-964 76434-966 76434-968 76435-008 76435-006 76435-128 76435-004 76435-126 76435-002 76435-124 76435-088 76435-000 76435-122 76435-086 76435-120 76435-084 76435-082 76435-080 76434-960 76434-962 76434-954 76434-956 76434-958 76435-118 76435-116 76435-114 76435-112 76435-110 76435-078 76435-076 76435-074 76435-192 76435-072 76435-190 76435-070 76434-950 76434-952 76434-942 76434-944 76434-946 76434-948 76435-108 76435-106 76435-104 76435-068 76435-102 76435-066 76435-188 76435-100 76435-064 76435-186 76435-062 76435-184 76435-060 76435-182 76435-180 76434-940 76431-176 76434-932 76431-174 76434-934 76431-172 76434-936 76434-938 76431-178 76435-178 76435-058 76435-176 76435-056 76435-174 76435-054 76435-172 76431-180 76435-052 76435-170 76435-050 76434-930 76434-920 76434-922 76434-924 76431-184 76434-926 76431-182 76434-928 76435-048 76430-422 76435-046 76430-424 76435-168 76435-044 76430-426 76435-166 76435-042 76435-164 76430-428 76435-040 76435-162 76435-160 76430-420 76434-998 76434-912 76434-914 76434-916 76430-418 76434-918 76430-410 76435-158 76435-038 76430-412 76435-156 76435-036 76430-414 76435-154 76435-034 76430-416 76435-152 76435-032 76435-150 76435-030 76434-990 76434-992 76434-994 76434-996 76434-986 76434-988 76435-028
Avantor® Hichrom HI-5, GC Columns
Chromatography Columns
Versatile, general purpose, low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 is a crossbonded phase and assure a long lifetime even under high temperature limits.
Max temperature limits may change depending on the film thickness.