Avantor® Hichrom HI-SE54, GC Columns
Supplier: VWR
Avantor® Hichrom
HI0532010105 HI05-10-010-10 HI05-10-010-15 HI05-32-025-50 HI05-53-265-10 HI05-20-010-60 HI05-53-100-75 HI05-05-005-2 HI0532025105 HI05-32-300-50 HI05-18-040-10 HI05-25-025-50 HI05-25-010-50 HI05-20-020-60 HI05-10-020-10 HI05-53-100-10 HI05-32-400-30 HI05-20-020-50 HI05-10-040-5 HI05-32-010-60 HI05-32-025-60 HI05-32-050-30 HI0525025105 HI05-32-150-30 HI05-53-300-75 HI05-18-018-10 HI05-20-100-10 HI05-05-005-5 HI05-25-100-15 HI05-53-100-30 HI05-18-018-20 HI05-53-025-30 HI05-20-020-20 HI05-18-018-40 HI05-15-015-15 HI05-18-020-20 HI05050052.5 HI05-53-700-30 HI05-25-050-15 HI0532150105 HI05-32-150-10 HI05-53-025-50 HI05-32-025-15 HI053210030G5 HI05-53-010-30 HI0532050105 HI05-32-100-60 HI05-25-050-60 HI05-53-150-75 HI0525100105 HI05-32-050-50 HI05-20-010-50 HI05-25-025-60 HI05-32-100-10 HI05-18-020-60 HI05-32-150-15 HI05-32-100-30 HI05-25-100-60 HI05-18-020-50 HI05-53-700-15 HI05-32-100-50 HI05-18-040-50 HI05-18-020-40 HI05-53-300-60 HI05-20-040-40 HI05-18-040-40 HI05-53-300-15 HI05-18-018-60 HI05-53-100-15 HI053201010G2 HI05-53-150-30 HI0532300105 HI05-32-010-50 HI0532100105 HI05-32-050-60 HI05-53-500-10 HI05-32-150-60 HI05-10-010-5 HI05-25-050-50 HI05-53-050-10 HI05-32-500-60 HI05-25-010-15 HI05-32-500-50 HI05-32-300-60 HI05-53-010-10 HI05-25-050-30 HI05-53-150-60 HI05-20-100-40 HI05-15-015-20 HI055305030G10 HI05-53-150-50 HI05-53-150-15 HI05320108G2 HI05-53-500-30 HI05-53-300-50 HI05-53-100-60 HI05-53-050-60 HI05-53-300-30 HI05-25-010-30 HI05-18-018-50 HI0532500105 HI05-18-020-10 HI05-20-010-20 HI052502515G5 HI05-32-010-15 HI05-10-040-15 HI05-32-300-15 HI05-20-010-40 HI05-53-050-15 HI05-25-010-60 HI05-25-100-50 HI05-32-025-10 HI05-32-300-30 HI05-18-040-20 HI05-53-025-15 HI05-10-020-15 HI05-32-025-30 HI05-25-100-30 HI05-20-020-10 HI05-15-015-5 HI05-15-015-30 HI05-53-010-15 HI05-20-040-60 HI05-32-100-15 HI05-53-500-75 HI05-10-040-10 HI05-20-100-20 HI05-32-050-15 HI05-53-500-60 HI05-53-150-10 HI05-53-100-50 HI05-32-010-10 HI05-25-025-15 HI05-25-100-10 HI05-15-015-10 HI05-20-100-60 HI052502560G5 HI05-53-050-50 HI05-32-500-30 HI05-53-500-15 HI05-10-020-5 HI05-32-300-10 HI05-20-020-40 HI05-20-010-10 HI05-25-025-10 HI05-53-300-10 HI05-20-040-20 HI05-18-040-60 HI052510030G5 HI052502530G5 HI05-32-150-50 HI05-20-040-50 HI05-25-025-30 HI05-25-050-10 HI05-20-040-10 HI05-32-010-30 HI0525050105 HI05-32-050-10 HI05-53-500-50 HI05-53-025-60 HI05-53-050-30 HI05-20-100-50 HI05-32-500-10 HI05-32-500-15 HI055310030G5
76435-664EA 1690.13 CAD
76435-664 76435-544 76435-662 76435-542 76431-408 76435-660 76435-540 76431-406 76431-416 76435-538 76431-414 76435-536 76435-658 76431-412 76435-534 76435-656 76431-410 76435-532 76435-654 76435-530 76435-498 76435-652 76435-496 76435-650 76431-418 76435-494 76435-492 76435-490 76430-946 76435-648 76435-528 76435-646 76435-526 76435-644 76431-420 76435-524 76435-488 76435-642 76435-522 76435-640 76435-486 76435-520 76435-484 76435-482 76435-480 76435-518 76435-516 76435-638 76435-514 76435-636 76435-512 76435-634 76435-510 76435-598 76435-632 76435-478 76435-596 76435-476 76435-630 76435-594 76435-474 76435-592 76435-472 76435-590 76435-628 76431-404 76435-508 76435-626 76435-748 76435-506 76435-624 76435-746 76435-504 76435-622 76435-744 76435-502 76435-620 76435-588 76435-742 76435-500 76435-586 76435-740 76430-920 76435-584 76430-922 76435-582 76435-580 76435-738 76430-914 76435-618 76435-736 76435-616 76430-916 76435-734 76430-918 76435-614 76435-732 76435-578 76435-612 76435-576 76435-730 76435-698 76435-610 76435-574 76435-696 76435-572 76435-694 76435-570 76435-692 76435-690 76435-608 76435-606 76435-728 76435-604 76435-726 76435-602 76435-724 76435-600 76435-688 76435-568 76435-722 76435-686 76435-720 76435-566 76430-940 76435-684 76435-564 76435-682 76430-942 76435-562 76435-680 76430-944 76435-560 76435-718 76435-716 76430-936 76435-714 76430-938 76435-558 76435-712 76435-710 76435-556 76435-678 76435-554 76435-676 76435-552 76435-674 76430-930 76435-550 76435-672 76430-932 76435-670 76430-934 76435-708 76430-924 76435-706 76430-926 76435-704 76435-668 76430-928 76435-548 76435-702 76435-666 76435-546 76435-700
Avantor® Hichrom HI-SE54, GC Columns
Chromatography Columns
General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, allergens, pesticides, and so on.
Max temperature limits may change depending on the film thickness.