Claritas PPT® Grade ICP-MS Tuning Solutions, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
Claritas PPT® Multi-Element Standards for ICP-MS. Tuning solutions are for ICP-MS instrumentation tuning and mass calibration prior to analysis.
The Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.
ICP-MS Tuning Solution 1; A dilution of 100-fold to 1,000-fold, depending on the sensitivity of the instrument, is suggested. Dilute with equal parts of Claritas PPT® Nitric Acid Blank and Water Blank to yield a 1% nitric acid matrix.
ICP-MS Tuning Solution 2: A dilution of 1,000-fold is suggested. Dilute with Claritas PPT® Nitric Acid Blank and Water Blank to yield a 1% nitric acid matrix
ICP-MS Tuning Solution 3: A dilution of 1,000-fold is suggested. Dilute with Claritas PPT® Nitric Acid Blank and Water Blank to yield a 0.5% nitric acid matrix.
ICP-MS Tuning Solution 4: A dilution of 100-fold to 1,000-fold is suggested. Dilute with Claritas PPT® Nitric Acid Blank to match your sample matrix."
Certifications: SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.