HotBlock® COD Reactor, Environmental Express®
Supplier: Environmental Express
B3000 B3000-240
76436-812EA 7753.85 CAD
76436-812 76436-814
HotBlock® COD Reactor, Environmental Express®
Digestion Systems Mineralisation Workstations
Digest two analytical batches at once!
  • Process up to fifty-six standard, 16-mm diameter, round-bottom COD vials
  • See actual and set-point temperatures simultaneously
  • Pre-programmed to run at 150 °C for two hours, and flashes when the cycle is complete
  • Maintain a setpoint of ±0.1 °C and sample consistency of ±1.5 °C
Reliable and uniform HotBlock technology now in a COD reactor. Process up to 56 samples, two analytical batches, in a single run! The digital display shows the actual temperature (in red) and set temperature (in green) simultaneously.