UV/VIS Photometric Accuracy Calibration Standards, FUV Dual Series, FireFlySci
Supplier: FIREFLYSCI, INC.
FUV-0.3D FUV-0.5D FUV-1.0D FUV-1.5D FUV-2.5D FUV-3.0D FUV-4.0D
76016-730EA 4078.42 CAD
76016-730 76016-732 76016-736 76016-738 76016-742 76016-744 76016-746
UV/VIS Photometric Accuracy Calibration Standards, FUV Dual Series, FireFlySci
Standards Colour Reference and Spectroscopy Standards
Firefly’s FUV Dual Series filters allow you to verify photometric accuracy and stray light in the range of 200 - 700 nm. Includes certified values for both the front and back of the filter for determining stray light.
  • Highly accurate and consistent spectrophotometer calibration every time
  • Never has to be replaced
  • Never has to be recalibrated if used properly
  • Scratch-resistant coating protects optical surfaces
  • Can be used in any spectrophotometer (with standard 12.5 x 12.5 x 45mm holder)
  • Surpasses NIST-2031 requirements and tolerances
Certifications: Certificate of calibration with NIST traceability.
Delivery information: Supplied with instruction manual.
Caution: Note: The FUV-0.2D and below are highly sensitive and only for use with customers with the following high-end machines: Cary-5000, Cary-7000, Lambda 900, Lambda 950, Lambda 1050 or Shimadzu UV-3600.