Anti-CD274 Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
600-101-GU8 600-101-GU8S
76309-332EA 834.24 CAD
76309-332 76309-334
Anti-CD274 Goat Polyclonal Antibody
Antibodies
PDL1 belongs to the immunoglobulin superfamily and BTN/MOG family. PDL1 is involved in the costimulatory signal, essential for T-cell proliferation and production of IL-10 and IFNG, in an IL-2 dependent and a PDCD1 independent manner. It is up-regulated in T-and B-cells, dendritic cells, keratinocytes and monocytes after LPS and IFNG activation. PDL1 is up-regulated in B-cells activated by surface Ig cross-linking. Interaction with PDCD1 inhibits T-cell proliferation and cytokine production. During infection or inflammation of normal tissue, this interaction is important for preventing autoimmunity by maintaining homeostasis of the immune response. In tumor microenvironments, this interaction provides an immune escape for tumor cells through cytotoxic T-cell inactivation. Expression of this gene in tumor cells is considered to be prognostic in many types of human malignancies, including colon cancer and renal cell carcinoma. Anti-PDL-1 Antibody is useful for researches interested in cancer research, NF-kB pathway research, and the Immune system.

Type: Primary
Antigen: CD274
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR