Echo® and Airo® Filtered Fume Hood Accessories, Labconco
Supplier: Labconco
9577400 9577401 9577460 9577421 9577422 9577423 9577424 9577433 9577434 9577460 9577461 9577473 9577474
10746-776EA 2543.8 CAD
10746-776 10769-428 76436-344 76436-346 76436-348 76436-350 76436-352 76436-354 76436-356 77918-396 77918-398 77918-400 77918-402
Echo® and Airo® Filtered Fume Hood Accessories, Labconco
Fume Hoods and Enclosures Ductless Fume Hoods
Required and replacement accessories for Protector filtered fume hoods. These accessories are for use with Labconco Protector Echo® and Airo® filtered fume hoods. For all filters and prefilters: The number of filters/prefilters required depends on the width of the hood and if it is an initial order or a replacement order.
Neutrodine Unisorb molecular carbon filter for multi-purpose use with acids/bases/solvents, separated or combined. Compatible with over 700 chemicals.
HEPA filter, 99.995% efficient, H14 type.
Replacement roughing prefilter, located in light box.
Replacement acid roughing prefilter, located in light box. Treated for use with acids.
Replacement ammonia roughing prefilter, located in light box. Treated for use with ammonia.
Acid gas sensor is required for proper hood function on models with acid sensor. Replacement is required after hood alarm alerts user that shelf-life has been met (approximately 2 years).
Formaldehyde gas sensor is required for proper hood function on models with formaldehyde sensor. Replacement is required after hood alarm alerts user that shelf-life has been met (approximately 2 years).