Edge® 48-919 Light-Duty Multi-Purpose Industrial Gloves, Ansell
Supplier: Ansell Healthcare
48919060 48919070 48919080 48919090 48919100 48919110
76424-284CS 1001.79 CAD
76424-284 76424-286 76424-288 76424-290 76424-292 76424-294
Edge® 48-919 Light-Duty Multi-Purpose Industrial Gloves, Ansell
Gloves
Cost effective solution for lightweight applications requiring a durable coating and secure grip in oily environments.
  • Excellent abrasion resistance
  • Double layer nitrile coating keeps hands dry and comfortable when handling oily parts and tools
  • Elastic knit-wrist ensures snug fit
  • Applicable in oily environments
Made of a seamless liner, fully dipped into NBR coating. The gloves are fully NBR dipped to provide protection against industrial fluid exposure. The rough palm coating ensures high secure grip in oily environment. Designed for workers whoMore Product Information