Edge® 48-706 Heavy-Duty Cut Resistant Industrial Gloves, Ansell
Supplier: Ansell Healthcare
48706060 48706070 48706080 48706090 48706100 48706110
76424-312CS 1919.66 CAD
76424-312 76424-314 76424-316 76424-318 76424-320 76424-322
Edge® 48-706 Heavy-Duty Cut Resistant Industrial Gloves, Ansell
Gloves
Cost effective solution for medium weight applications requiring a high level of cut performance in dry or slightly oily environments.
  • High durability Nitrile foam coating
  • Excellent comfort and dexterity
  • High level of cut performance EN ISO level C cut performance (ANSI Level 3)
  • Excellent abrasion resistance
  • Breathable liner provides a high level of comfort for end users
  • Good level of grip in dry and slightly oily environments
Made of a seamless liner, palm dipped into foam nitrile coating. ANSI cut level A3 and EN ISO 13997 rated level C provide aMore Product Information