Eisco® Safety Pack Glassware
Supplier: Wards
CH196031 CH196032 CH196033 CH196034 CH196041 CH196042 CH196046 CH196047 CH196048 CH196061
470328-044KT 37.75 CAD
470328-044 470328-046 470328-048 470328-050 470328-052 470328-054 470328-056 470328-058 470328-060 470328-062
Eisco® Safety Pack Glassware
Labware Sets
Convenient sets of glassware expertly packaged custom-fit foam inserts for worry-free transport and safe storage.
  • Convenient sets of high-quality Borosilicate 3.3 glassware
  • Packaged in thick, custom-fit foam inserts for safe transport and storage
  • Superior heat tolerance and chemical resistance
  • Autoclavable at 121 °C
  • Volumetric glassware is hand calibrated in Eisco's ISO 17025 compliant laboratory
Eisco safety pack glassware are all made of high-quality Borosilicate 3.3 glass. You will find these convenient sets of glassware expertly packaged in heavy duty boxes with thick, custom-fit foam inserts for worry-free transport and safe storage. All Eisco glassware pieces feature low temperature gradients and can withstand high temperatures as well as thermal shocks. They are microwaveable, autoclavable, and have superior chemical durability. Easy to read graduations are printed in white and are chemical resistant.
Eisco labs Borosilicate 3.3 glass has the following heat tolerances: Strain point −515 °C, annealing point 565 °C, softening point 820 °C.
Certifications: Eisco safety pack glassware is manufactured and calibrated to the following standards: Beakers - DIN 12231 and ISO 3819; Conical Flasks - DIN ISO 1773; Volumetric Flasks - ASTM-E288; Graduated Cylinders - ASTM-E1272.