SP Wilmad-LabGlass Large Writing Area High-Throughput NMR Tubes, 5 mm O.D., SP Industries
Supplier: SP Industries
WG-1003-7-25 WG-1003-7-50 WG-1003-7
76420-858PK 89.86 CAD
76420-858 76420-860 76420-862
SP Wilmad-LabGlass Large Writing Area High-Throughput NMR Tubes, 5 mm O.D., SP Industries
Tubes NMR Tubes
Double the writing area allows for easy sample marking and identification.
  • 2x larger marking area eliminates cramped and unreadable sample names
  • Silkscreened writing area keeps sample names from easily wiping off
  • Designed for routine use in most low to mid field NMR spectrometers
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimensions
These High-Throughput tubes have an average camber of 60 µm to ensure spectrum clarity for smallMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable