VWR® 96-Well PP Microplates
Supplier: VWR International
76397-816 76397-818 76397-574 76397-576 76397-578 76397-582
76397-816PK 150.64 CAD
76397-816 76397-818 76397-574 76397-576 76397-578 76397-582
VWR® 96-Well PP Microplates
Microplates Deep Well Microplates
VWR® 96-Well plates and covers are designed for chromatography and high throughput screening (HTS) applications.
  • SBS and ANSI standard footprint
  • Low binding virgin PP for homogenous assays and storage
  • –80 to +121 °C tolerant
VWR® 96-Well Microplates are manufactured specifically for chromatography. Multiple volumes and interior finishes allow for enhanced sample recovery. Low binding virgin polypropylene plates are perfect for high throughput screening (HTS), and sensitive samples.
Order Now
SPECIFICATIONS
Environmentally Preferable